Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-hydroxy-5-nitrobenzaldehyde, 95%

$80.61 - $212.46

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-hydroxy-5-nitrobenzaldehyde
CAS 711022-47-8
Molecular Formula C7H4FNO4
Formula Weight 185.11
MDL Number MFCD11846408
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5187803

  SDS
alfa aesar™
H5187803
1g Each for $212.46

AAH51878MD

  SDS
alfa aesar™
H51878MD
250mg Each for $80.61
Description & Specifications

Specifications

Formula Weight 185.11
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H4FNO4
Assay 95%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 1 Warning
Chemical Name or Material 3-Fluoro-4-hydroxy-5-nitrobenzaldehyde
CAS 711022-47-8
MDL Number MFCD11846408
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.