missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Fluoro-4-formylphenylboronic acid, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445600050
Specifications
3-Fluoro-4-formylphenylboronic acid | |
248270-25-9 | |
C7H6BFO3 | |
NZNRMUVHUVCIBR-UHFFFAOYSA-N | |
(3-fluoro-4-formylphenyl)boronic acid | |
2778654 | |
95% |
5g | |
94% min. (HPLC) | |
3-fluoranyl-4-methanoyl-phenyl boronic acid, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenboronic acid, 3-fluoro-4-formylphenyl boronic acid, 4-borono-3-fluorobenzaldehyde, acmc-1cga0, boronic acid, 3-fluoro-4-formylphenyl, ksc494k4h, pubchem1847 | |
B(C1=CC(=C(C=C1)C=O)F)(O)O | |
167.93 | |
167.93 |
Chemical Identifiers
248270-25-9 | |
167.93 | |
3-fluoranyl-4-methanoyl-phenyl boronic acid, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenboronic acid, 3-fluoro-4-formylphenyl boronic acid, 4-borono-3-fluorobenzaldehyde, acmc-1cga0, boronic acid, 3-fluoro-4-formylphenyl, ksc494k4h, pubchem1847 | |
(3-fluoro-4-formylphenyl)boronic acid |
C7H6BFO3 | |
NZNRMUVHUVCIBR-UHFFFAOYSA-N | |
2778654 | |
B(C1=CC(=C(C=C1)C=O)F)(O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning