missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-4-formylphenylboronic acid, 95%, ACROS Organics™

Chemical Identifiers

CAS 248270-25-9
Molecular Formula C7H6BFO3
Molecular Weight (g/mol) 167.93
InChI Key NZNRMUVHUVCIBR-UHFFFAOYSA-N
Synonym 3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenyl boronic acid, 3-fluoro-4-formylphenboronic acid, boronic acid, 3-fluoro-4-formylphenyl, pubchem1847, acmc-1cga0, ksc494k4h, 4-borono-3-fluorobenzaldehyde, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoranyl-4-methanoyl-phenyl boronic acid
PubChem CID 2778654
IUPAC Name (3-fluoro-4-formylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C=O)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445600050 SDS View Documents Acros Organics
AC445600050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445600010 SDS View Documents Acros Organics
AC445600010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

248270-25-9
167.93
3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenyl boronic acid, 3-fluoro-4-formylphenboronic acid, boronic acid, 3-fluoro-4-formylphenyl, pubchem1847, acmc-1cga0, ksc494k4h, 4-borono-3-fluorobenzaldehyde, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoranyl-4-methanoyl-phenyl boronic acid
(3-fluoro-4-formylphenyl)boronic acid
C7H6BFO3
NZNRMUVHUVCIBR-UHFFFAOYSA-N
2778654
B(C1=CC(=C(C=C1)C=O)F)(O)O

Specifications

5g
95%
3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenyl boronic acid, 3-fluoro-4-formylphenboronic acid, boronic acid, 3-fluoro-4-formylphenyl, pubchem1847, acmc-1cga0, ksc494k4h, 4-borono-3-fluorobenzaldehyde, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoranyl-4-methanoyl-phenyl boronic acid
B(C1=CC(=C(C=C1)C=O)F)(O)O
167.93
167.93
248270-25-9
C7H6BFO3
NZNRMUVHUVCIBR-UHFFFAOYSA-N
(3-fluoro-4-formylphenyl)boronic acid
2778654
3-Fluoro-4-formylphenylboronic acid
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Warning