Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-formylbenzeneboronic acid, 98%

$56.81 - $56.81

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-formylbenzeneboronic acid
CAS 248270-25-9
Molecular Formula C7H6BFO3
Formula Weight 167.93
MDL Number MFCD02093051
Synonym 4-Borono-3-fluorobenzaldehyde; 3-Fluoro-4-formylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1785103

  SDS
alfa aesar™
L1785103
1g Each for $56.81
Description & Specifications

Specifications

Formula Weight 167.93
Health Hazard 1 Warning
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 4-Borono-3-fluorobenzaldehyde; 3-Fluoro-4-formylphenylboronic acid
Quantity 1g
Chemical Name or Material 3-Fluoro-4-formylbenzeneboronic acid
Molecular Formula C7H6BFO3
CAS 248270-25-9
Melting Point ≈240°C
MDL Number MFCD02093051
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.