missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H62530MD

 View more versions of this product

Catalog No. AAH62530MD



Specifications

Specifications

3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
250mg
C17H25BFNO3
3-fluoro-4-morpholin-4-ylmethyl benzene boronic acid, pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester, 3-fluoro-4-n-morpholinomethyl phenylboronic acid pinacol ester, 3-fluoro-4-n-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl morpholine, 4-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CN3CCOCC3)F
321.199
321.19
96%
35°C to 40°C
1073354-74-1
MFCD09027076
QINFDPZSZHPUSC-UHFFFAOYSA-N
4-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
43811063
Solid

Chemical Identifiers

1073354-74-1
321.199
QINFDPZSZHPUSC-UHFFFAOYSA-N
43811063
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CN3CCOCC3)F
C17H25BFNO3
MFCD09027076
3-fluoro-4-morpholin-4-ylmethyl benzene boronic acid, pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester, 3-fluoro-4-n-morpholinomethyl phenylboronic acid pinacol ester, 3-fluoro-4-n-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl morpholine, 4-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine
4-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents