missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(4-morpholinylcarbonyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54607MD

 View more versions of this product

Catalog No. AAH54607MD



Specifications

Specifications

3-Fluoro-4-(4-morpholinylcarbonyl)benzeneboronic acid
1008119-70-7
MFCD20265298
PCOQCLUJTVPMSY-UHFFFAOYSA-N
[3-fluoro-4-(morpholine-4-carbonyl)phenyl]boronic acid
57618415
97%
250mg
C11H13BFNO4
3-fluoro-4-4-morpholinylcarbonyl benzeneboronic acid
B(C1=CC(=C(C=C1)C(=O)N2CCOCC2)F)(O)O
253.036
253.04

Chemical Identifiers

1008119-70-7
253.036
PCOQCLUJTVPMSY-UHFFFAOYSA-N
57618415
B(C1=CC(=C(C=C1)C(=O)N2CCOCC2)F)(O)O
C11H13BFNO4
MFCD20265298
3-fluoro-4-4-morpholinylcarbonyl benzeneboronic acid
[3-fluoro-4-(morpholine-4-carbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents