missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(4-methyl-1-piperidinylcarbonyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5433103

 View more versions of this product

Catalog No. AAH5433103



Description

Description

Specifications

Specifications

3-Fluoro-4-(4-methyl-1-piperidinylcarbonyl)benzeneboronic acid
1449132-53-9
MFCD20265245
NTDBICSBLSVBSL-UHFFFAOYSA-N
[3-fluoro-4-(4-methylpiperidine-1-carbonyl)phenyl]boronic acid
73995758
97%
1g
C13H17BFNO3
3-fluoro-4-4-methyl-1-piperidinylcarbonyl benzeneboronic acid, 3-fluoro-4-4-methyl-1-piperidinylcarbonyl phenylboronic acid, 3-fluoro-4-4-methylpiperidine-1-carbonyl phenyl boronic acid, 3-fluoro-4-4-methylpiperidine-1-carbonyl phenylboronic acid
B(C1=CC(=C(C=C1)C(=O)N2CCC(CC2)C)F)(O)O
265.091
265.09

Chemical Identifiers

1449132-53-9
265.091
NTDBICSBLSVBSL-UHFFFAOYSA-N
73995758
B(C1=CC(=C(C=C1)C(=O)N2CCC(CC2)C)F)(O)O
C13H17BFNO3
MFCD20265245
3-fluoro-4-4-methyl-1-piperidinylcarbonyl benzeneboronic acid, 3-fluoro-4-4-methyl-1-piperidinylcarbonyl phenylboronic acid, 3-fluoro-4-4-methylpiperidine-1-carbonyl phenyl boronic acid, 3-fluoro-4-4-methylpiperidine-1-carbonyl phenylboronic acid
[3-fluoro-4-(4-methylpiperidine-1-carbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents