Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-(trifluoromethyl)-DL-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-2-(trifluoromethyl)-DL-phenylalanine
Molecular Formula C10H9F4NO2
Formula Weight 251.18
MDL Number MFCD09832252
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH31874MD

 
alfa aesar™
H31874MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.18
Chemical Name or Material 3-Fluoro-2-(trifluoromethyl)-DL-phenylalanine
Assay 97%
Molecular Formula C10H9F4NO2
MDL Number MFCD09832252
Melting Point ≈251°C
Quantity 250mg