Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-(trifluoromethyl)benzoic acid, 97%

$64.19 - $158.54

Chemical Identifiers

Chemical Name or Material 3-Fluoro-2-(trifluoromethyl)benzoic acid
CAS 261951-80-8
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD01631565
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2673203

 
alfa aesar™
H2673203
1g Each for $158.54

AAH26732MD

 
alfa aesar™
H26732MD
250mg Each for $64.19
Description & Specifications

Specifications

Formula Weight 208.11
Chemical Name or Material 3-Fluoro-2-(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C8H4F4O2
MDL Number MFCD01631565
CAS 261951-80-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 118° to 121°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.