Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-(trifluoromethyl)benzaldehyde, 97%

$67.05 - $67.05

Chemical Identifiers

Chemical Name or Material 3-Fluoro-2-(trifluoromethyl)benzaldehyde
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD04116354
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH32180MD

  SDS
alfa aesar™
H32180MD
250mg Each for $67.05

AAH3218003

  SDS
alfa aesar™
H3218003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-Fluoro-2-(trifluoromethyl)benzaldehyde
Quantity 250mg
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD04116354
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.