missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Fluoro-2-pyridinecarboxaldehyde, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC448620010
Specifications
3-Fluoro-2-pyridinecarboxaldehyde | |
51.0°C to 54.0°C | |
31224-43-8 | |
C6H4FNO | |
OZIMPUNGBUYCSP-UHFFFAOYSA-N | |
3-fluoropyridine-2-carbaldehyde | |
11344017 | |
97% |
Glass bottle | |
1g | |
96% min. (GC) | |
2-formyl-3-fluoropyridine, 2-pyridinecarboxaldehyde, 3-fluoro, 3-fluoro-2-formyl-pyridine, 3-fluoro-2-formylpyridine, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 3-fluoropicolinaldehyde, 3-fluoropyridine-2-carboxaldehyde, acmc-209hkw, pubchem5142 | |
C1=CC(=C(N=C1)C=O)F | |
125.1 | |
125.1 |
Chemical Identifiers
31224-43-8 | |
125.1 | |
2-formyl-3-fluoropyridine, 2-pyridinecarboxaldehyde, 3-fluoro, 3-fluoro-2-formyl-pyridine, 3-fluoro-2-formylpyridine, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 3-fluoropicolinaldehyde, 3-fluoropyridine-2-carboxaldehyde, acmc-209hkw, pubchem5142 | |
3-fluoropyridine-2-carbaldehyde |
C6H4FNO | |
OZIMPUNGBUYCSP-UHFFFAOYSA-N | |
11344017 | |
C1=CC(=C(N=C1)C=O)F |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning