Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-(phenoxycarbonylamino)benzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-2-(phenoxycarbonylamino)benzeneboronic acid pinacol ester
Formula Weight 357.18
Synonym 3-Fluoro-2-(phenoxycarbonylamino)phenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6201603

 
alfa aesar™
H6201603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62016MD

 
alfa aesar™
H62016MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 357.18
Synonym 3-Fluoro-2-(phenoxycarbonylamino)phenylboronic acid pinacol ester
Quantity 1g
Chemical Name or Material 3-Fluoro-2-(phenoxycarbonylamino)benzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%