missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-nitrotoluene, 96%

Manufacturer:  Alfa Aesar™ H6357406

 View more versions of this product

Catalog No. AAH6357406



Specifications

Specifications

3-Fluoro-2-nitrotoluene
17°C to 18°C
3013-27-2
MFCD08458093
3-fluoro-2-nitrotoluene, 1-fluoro-3-methyl-2-nitro-benzene, 2-fluoro-6-methylnitrobenzene, benzene, 1-fluoro-3-methyl-2-nitro, fluoronitrotoluene, fluor-nitro-toluole, pubchem18581, 2-nitro-3-fluorotoluene, ksc497k3l, 3-methyl-2-nitrofluorobenzene
CC1=C(C(=CC=C1)F)[N+](=O)[O-]
155.128
155.13
96%
White
5g
C7H6FNO2
UN2811
JSXAJLMUBSEJFF-UHFFFAOYSA-N
1-fluoro-3-methyl-2-nitrobenzene
19773803
Low Melting Solid

Chemical Identifiers

3013-27-2
155.128
JSXAJLMUBSEJFF-UHFFFAOYSA-N
19773803
CC1=C(C(=CC=C1)F)[N+](=O)[O-]
C7H6FNO2
MFCD08458093
3-fluoro-2-nitrotoluene, 1-fluoro-3-methyl-2-nitro-benzene, 2-fluoro-6-methylnitrobenzene, benzene, 1-fluoro-3-methyl-2-nitro, fluoronitrotoluene, fluor-nitro-toluole, pubchem18581, 2-nitro-3-fluorotoluene, ksc497k3l, 3-methyl-2-nitrofluorobenzene
1-fluoro-3-methyl-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C

SDS Documents