Learn More
3-Fluoro-2-methylpyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6470503
Description
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
3-Fluoro-2-methylpyridine | |
Colorless to Yellow | |
MFCD08705710 | |
6497832 | |
Slightly soluble in water. | |
CC1=C(C=CC=N1)F | |
111.119 | |
111.12 | |
Liquid |
15931-15-4 | |
C6H6FN | |
1 g | |
3-fluoro-2-picoline, pyridine, 3-fluoro-2-methyl, 2-methyl-3-fluoropyridine, 3-fluoro-2-methyl pyridine, 3-fluoro-2-methyl-pyridine, fluoropicoline, pubchem14381, acmc-209xci, ksc530a3b, pyridine,3-fluoro-2-methyl | |
IRIHSZDBTACXCT-UHFFFAOYSA-N | |
3-fluoro-2-methylpyridine | |
21613172 | |
98% |
Chemical Identifiers
15931-15-4 | |
111.119 | |
IRIHSZDBTACXCT-UHFFFAOYSA-N | |
21613172 | |
CC1=C(C=CC=N1)F |
C6H6FN | |
MFCD08705710 | |
3-fluoro-2-picoline, pyridine, 3-fluoro-2-methyl, 2-methyl-3-fluoropyridine, 3-fluoro-2-methyl pyridine, 3-fluoro-2-methyl-pyridine, fluoropicoline, pubchem14381, acmc-209xci, ksc530a3b, pyridine,3-fluoro-2-methyl | |
3-fluoro-2-methylpyridine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only