Learn More
Alfa Aesar™ 3-Fluoro-2-methylaniline, 98+%
$79.09 - $284.32
Chemical Identifiers
CAS | 443-86-7 |
---|---|
Molecular Formula | C7H8FN |
Molecular Weight (g/mol) | 125.146 |
MDL Number | MFCD00007760 |
InChI Key | SLDLVGFPFFLYBM-UHFFFAOYSA-N |
Synonym | 2-amino-6-fluorotoluene, 2-fluoro-6-aminotoluene, 2-methyl-3-fluoroaniline, 3-fluor-2-methylanilin, 3-fluoro-2-methyl-aniline, 3-fluoro-2-methylphenylamine, 3-fluoro-o-toluidine, 6-amino-2-fluorotoluene, benzenamine, 3-fluoro-2-methyl, hydrochloride |
PubChem CID | 285553 |
IUPAC Name | 3-fluoro-2-methylaniline |
SMILES | CC1=C(C=CC=C1F)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2276406
|
Alfa Aesar™
B2276406 |
5g |
Each for $79.09
|
|
AAB2276414
|
Alfa Aesar™
B2276414 |
25g |
Each for $284.32
|
|
Chemical Identifiers
443-86-7 | |
125.146 | |
SLDLVGFPFFLYBM-UHFFFAOYSA-N | |
285553 | |
CC1=C(C=CC=C1F)N |
C7H8FN | |
MFCD00007760 | |
2-amino-6-fluorotoluene, 2-fluoro-6-aminotoluene, 2-methyl-3-fluoroaniline, 3-fluor-2-methylanilin, 3-fluoro-2-methyl-aniline, 3-fluoro-2-methylphenylamine, 3-fluoro-o-toluidine, 6-amino-2-fluorotoluene, benzenamine, 3-fluoro-2-methyl, hydrochloride | |
3-fluoro-2-methylaniline |
Specifications
1.099 | |
Characteristic | |
205°C | |
5g | |
C7H8FN | |
UN2810 | |
SLDLVGFPFFLYBM-UHFFFAOYSA-N | |
3-fluoro-2-methylaniline | |
285553 | |
≥98% |
86°C (186°F) | |
1.542 | |
7°C | |
443-86-7 | |
MFCD00007760 | |
2-amino-6-fluorotoluene, 2-fluoro-6-aminotoluene, 2-methyl-3-fluoroaniline, 3-fluor-2-methylanilin, 3-fluoro-2-methyl-aniline, 3-fluoro-2-methylphenylamine, 3-fluoro-o-toluidine, 6-amino-2-fluorotoluene, benzenamine, 3-fluoro-2-methyl, hydrochloride | |
CC1=C(C=CC=C1F)N | |
125.146 | |
125.15 | |
3-Fluoro-2-methylaniline |
Safety and Handling
GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
H227-H301+H311+H331-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 207-142-4
TSCA : No
Recommended Storage : Ambient temperatures