Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Fluoro-2'-methylacetophenone, 97%

$121.46 - $483.34

Chemical Identifiers

Chemical Name or Material 3'-Fluoro-2'-methylacetophenone
CAS 177942-47-1
Molecular Formula C9H9FO
Formula Weight 152.17
MDL Number MFCD09832257
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3222103

 
alfa aesar™
H3222103
1g Each for $121.46

AAH3222106

 
alfa aesar™
H3222106
5g Each for $483.34
Description & Specifications

Specifications

Formula Weight 152.17
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 1g
Chemical Name or Material 3'-Fluoro-2'-methylacetophenone
Assay 97%
Molecular Formula C9H9FO
MDL Number MFCD09832257
CAS 177942-47-1
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.