Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-methoxybenzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-2-methoxybenzonitrile
Molecular Formula C8H6FNO
Formula Weight 151.14
MDL Number MFCD06637312
UN Number UN3276
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3228403

  SDS
alfa aesar™
H3228403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3228406

  SDS
alfa aesar™
H3228406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.14
Assay 97%
Molecular Formula C8H6FNO
Chemical Name or Material 3-Fluoro-2-methoxybenzonitrile
MDL Number MFCD06637312
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Danger
UN Number UN3276
Quantity 1g
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.