missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-methoxybenzoic acid, 98%

$182.50 - $459.24

Chemical Identifiers

CAS 106428-05-1
Molecular Formula C8H7FO3
Formula Weight 170.14
MDL Number MFCD00973972
Chemical Name or Material 3-Fluoro-2-methoxybenzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2616404 SDS Alfa Aesar™
H2616404
2g Each for $459.24
AAH26164ME SDS Alfa Aesar™
H26164ME
500mg Each for $182.50
Specifications Description & Specifications

Chemical Identifiers

106428-05-1
170.14
3-Fluoro-2-methoxybenzoic acid
C8H7FO3
MFCD00973972

Specifications

112° to 114°C
106428-05-1
170.14
3-Fluoro-2-methoxybenzoic acid
2g
C8H7FO3
MFCD00973972
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning