Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-methoxybenzaldehyde, 96%

$42.04 - $174.78

Chemical Identifiers

Chemical Name or Material 3-Fluoro-2-methoxybenzaldehyde
CAS 74266-68-5
Molecular Formula C8H7FO2
Formula Weight 154.14
MDL Number MFCD04112486
Synonym 3-Fluoro-o-anisaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3297103

  SDS
alfa aesar™
H3297103
1g Each for $42.04

AAH3297106

  SDS
alfa aesar™
H3297106
5g Each for $174.78
Description & Specifications

Specifications

Formula Weight 154.14
Assay 96%
Health Hazard 1 Warning
CAS 74266-68-5
MDL Number MFCD04112486
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3-Fluoro-2-methoxybenzaldehyde
Molecular Formula C8H7FO2
Synonym 3-Fluoro-o-anisaldehyde
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.