missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-hydroxybenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Synonym 3-Fluoro-2-hydroxyphenylboronic acid pinacol ester
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62559MD
View Documents
Alfa Aesar™
H62559MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6255903
View Documents
Alfa Aesar™
H6255903
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

3-Fluoro-2-hydroxyphenylboronic acid pinacol ester

Specifications

250mg
238.06
96%
3-Fluoro-2-hydroxyphenylboronic acid pinacol ester
Solid
3-Fluoro-2-hydroxybenzeneboronic acid pinacol ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C