missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-2-hydroxybenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H62559MD

 View more versions of this product

Catalog No. AAH62559MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Fluoro-2-hydroxybenzeneboronic acid pinacol ester
3-Fluoro-2-hydroxyphenylboronic acid pinacol ester
Solid
250mg
238.06
96%

Chemical Identifiers

3-Fluoro-2-hydroxyphenylboronic acid pinacol ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

Documents