missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-exo-Aminobicyclo(2.2.1)heptane-2-exo-carboxylic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 265680010
Specifications
3-exo-Aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid | |
Glass Bottle | |
1g | |
88330-32-9 | |
100.0 | |
C8H13NO2 | |
1alpha,4alpha-3alpha-aminobicyclo 2.2.1 heptane-2alpha-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 heptane-2-carboxylic acid, 3-exo-aminobicyclo 2.2.1 heptane-2-carboxylic acid, bicyclo 2.2.1 heptane-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci | |
C1CC2CC1C(C2N)C(=O)O | |
155.197 | |
155.2 | |
≥97.5% (on dry substance) |
Authentic | |
White | |
98% | |
97.5 | |
98% | |
MFCD00143159 | |
JSYLGUSANAWARQ-KBGZMJGOSA-N | |
(2R,3S)-3-aminobicyclo[2.2.1]heptane-2-carboxylic acid | |
59451722 | |
Crystalline Powder |
Chemical Identifiers
88330-32-9 | |
155.197 | |
JSYLGUSANAWARQ-KBGZMJGOSA-N | |
59451722 | |
C1CC2CC1C(C2N)C(=O)O |
C8H13NO2 | |
MFCD00143159 | |
1alpha,4alpha-3alpha-aminobicyclo 2.2.1 heptane-2alpha-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 heptane-2-carboxylic acid, 3-exo-aminobicyclo 2.2.1 heptane-2-carboxylic acid, bicyclo 2.2.1 heptane-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci | |
(2R,3S)-3-aminobicyclo[2.2.1]heptane-2-carboxylic acid |
Safety and Handling
TSCA : TSCA