missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-exo-Aminobicyclo(2.2.1)heptane-2-exo-carboxylic acid, 98%, ACROS Organics™
$778.51 - $778.51
Chemical Identifiers
CAS | 88330-32-9 |
---|---|
Molecular Formula | C8H13NO2 |
Molecular Weight (g/mol) | 155.197 |
MDL Number | MFCD00143159 |
InChI Key | JSYLGUSANAWARQ-KBGZMJGOSA-N |
Synonym | 1alpha,4alpha-3alpha-aminobicyclo 2.2.1 heptane-2alpha-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 heptane-2-carboxylic acid, 3-exo-aminobicyclo 2.2.1 heptane-2-carboxylic acid, bicyclo 2.2.1 heptane-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci |
PubChem CID | 59451722 |
IUPAC Name | (2R,3S)-3-aminobicyclo[2.2.1]heptane-2-carboxylic acid |
SMILES | C1CC2CC1C(C2N)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC265680010
|
Thermo Fisher Scientific
265680010 |
1g | Glass Bottle |
Each for $778.51
|
|
Chemical Identifiers
88330-32-9 | |
155.197 | |
JSYLGUSANAWARQ-KBGZMJGOSA-N | |
59451722 | |
C1CC2CC1C(C2N)C(=O)O |
C8H13NO2 | |
MFCD00143159 | |
1alpha,4alpha-3alpha-aminobicyclo 2.2.1 heptane-2alpha-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 heptane-2-carboxylic acid, 3-exo-aminobicyclo 2.2.1 heptane-2-carboxylic acid, bicyclo 2.2.1 heptane-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci | |
(2R,3S)-3-aminobicyclo[2.2.1]heptane-2-carboxylic acid |
Specifications
Authentic | |
White | |
88330-32-9 | |
100.0 | |
C8H13NO2 | |
1alpha,4alpha-3alpha-aminobicyclo 2.2.1 heptane-2alpha-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 heptane-2-carboxylic acid, 3-exo-aminobicyclo 2.2.1 heptane-2-carboxylic acid, bicyclo 2.2.1 heptane-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci | |
C1CC2CC1C(C2N)C(=O)O | |
155.197 | |
155.2 | |
≥97.5% (on dry substance) |
Glass Bottle | |
1g | |
97.5 | |
98% | |
MFCD00143159 | |
JSYLGUSANAWARQ-KBGZMJGOSA-N | |
(2R,3S)-3-aminobicyclo[2.2.1]heptane-2-carboxylic acid | |
59451722 | |
Crystalline Powder | |
3-exo-Aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid, 98% |
Safety and Handling
TSCA : TSCA