missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%, ACROS Organics™
Chemical Identifiers
CAS |
92511-32-5 |
Molecular Formula |
C8H11NO2 |
Molecular Weight (g/mol) |
153.18 |
MDL Number |
MFCD00143158 |
InChI Key |
FCYFJGGJCJDCPB-BNHYGAARSA-N |
Synonym |
1r,1beta,4s,4beta-3alpha-amino-2alpha-norborna-5-enecarboxylic acid, 1r,2r,3s,4s/1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 1r,2s,3 r,4s-3-amino-5-bicyclo 2,2,1 heptene-2-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-endo-aminobicyclo 2.2.1 hept-5-ene-2-endo-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci, bicyclo 2.2.1 hept-5-ene-2-carboxylicacid, 3-amino-, 1r,2s,3r,4s, diendo-3-amino-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, rac-1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid |
PubChem CID |
13160880 |
IUPAC Name |
(1R,2S,3R,4S)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid |
SMILES |
C1C2C=CC1C(C2C(=O)O)N |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
92511-32-5 |
|
153.18 |
|
FCYFJGGJCJDCPB-BNHYGAARSA-N |
|
13160880 |
|
C1C2C=CC1C(C2C(=O)O)N |
|
C8H11NO2 |
|
MFCD00143158 |
|
1r,1beta,4s,4beta-3alpha-amino-2alpha-norborna-5-enecarboxylic acid, 1r,2r,3s,4s/1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 1r,2s,3 r,4s-3-amino-5-bicyclo 2,2,1 heptene-2-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-endo-aminobicyclo 2.2.1 hept-5-ene-2-endo-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci, bicyclo 2.2.1 hept-5-ene-2-carboxylicacid, 3-amino-, 1r,2s,3r,4s, diendo-3-amino-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, rac-1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid |
|
(1R,2S,3R,4S)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid |
Specifications
|
Authentic |
|
10.5% max. (K.F.) |
|
261.0°C to 263.0°C |
|
92511-32-5 |
|
100.0 |
|
C8H11NO2 |
|
1r,1beta,4s,4beta-3alpha-amino-2alpha-norborna-5-enecarboxylic acid, 1r,2r,3s,4s/1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 1r,2s,3 r,4s-3-amino-5-bicyclo 2,2,1 heptene-2-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-endo-aminobicyclo 2.2.1 hept-5-ene-2-endo-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci, bicyclo 2.2.1 hept-5-ene-2-carboxylicacid, 3-amino-, 1r,2s,3r,4s, diendo-3-amino-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, rac-1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid |
|
FCYFJGGJCJDCPB-BNHYGAARSA-N |
|
(1R,2S,3R,4S)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid |
|
13160880 |
|
Powder |
|
3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid, 98% |
|
Glass bottle |
|
Beige to Gray |
|
1g |
|
97.5 |
|
98% |
|
MFCD00143158 |
|
Solubility in water: soluble. Other solubilities: soluble in ethanol and methanol |
|
C1C2C=CC1C(C2C(=O)O)N |
|
153.18 |
|
153.18 |
|
98% |
Safety and Handling
SDS