missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 277812500

 View more versions of this product

Catalog No. AC277812500



Specifications

Specifications

3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide
Glass bottle
235.0°C to 253.0°C
99+%
99.0
99+%
MFCD00143157
Solubility in water: .. Other solubilities: soluble in ethanol and methanol
C1C2C=CC1C(C2C(=O)N)N
152.2
152.2
99+%
Authentic
White
250mg
105786-40-1
100.0
C8H12N2O
+-1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, +/--1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, bicyclo 2.2.1 hept-5-ene-2-carboxamide, 3-amino-, 1r,2r,3s,4s-rel-9ci
SCQSHSJVMGGQKR-JRTVQGFMSA-N
(1S,2S,3R,4R)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxamide
42580098
Crystalline Powder

Chemical Identifiers

105786-40-1
152.2
SCQSHSJVMGGQKR-JRTVQGFMSA-N
42580098
C1C2C=CC1C(C2C(=O)N)N
C8H12N2O
MFCD00143157
+-1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, +/--1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, bicyclo 2.2.1 hept-5-ene-2-carboxamide, 3-amino-, 1r,2r,3s,4s-rel-9ci
(1S,2S,3R,4R)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxamide
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents