missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethynyltoluene, TCI America™

Manufacturer:  TCI America E066525G

 View more versions of this product

Catalog No. E066525G



Specifications

Specifications

3-Ethynyltoluene
Yellow
766-82-5
3295
RENYIDZOAFFNHC-UHFFFAOYSA-N
1-ethynyl-3-methylbenzene
136600
Clear Liquid at 20°C
60°C
25g
C9H8
3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
CC1=CC=CC(=C1)C#C
116.163
116.16
≥98.0% (GC)

Chemical Identifiers

766-82-5
116.163
3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
1-ethynyl-3-methylbenzene
C9H8
RENYIDZOAFFNHC-UHFFFAOYSA-N
136600
CC1=CC=CC(=C1)C#C
Safety and Handling

Safety and Handling

TSCA : No

Documents