Learn More
3-Ethynylthiophene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5589806
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications
3-Ethynylthiophene | |
1.098 | |
46°C (115°F) | |
1.58 | |
5 g | |
3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b | |
MJHLPKWONJUCFK-UHFFFAOYSA-N | |
3-ethynylthiophene | |
3548422 | |
96% |
67237-53-0 | |
152°C to 153°C | |
C6H4S | |
MFCD04039973 | |
UN1993 | |
Slightly soluble in water. | |
C#CC1=CSC=C1 | |
108.158 | |
108.16 |
Chemical Identifiers
67237-53-0 | |
108.158 | |
MJHLPKWONJUCFK-UHFFFAOYSA-N | |
3548422 | |
C#CC1=CSC=C1 |
C6H4S | |
MFCD04039973 | |
3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b | |
3-ethynylthiophene |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H318-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only