missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethynylpyridine, 97%

Manufacturer:  Alfa Aesar™ H5346506

 View more versions of this product

Catalog No. AAH5346506



Specifications

Specifications

3-Ethynylpyridine
83°C to 84°C (30mmHg)
5g
C7H5N
UN1325
CLRPXACRDTXENY-UHFFFAOYSA-N
3-ethynylpyridine
186003
97%
57°C (135°F)
39°C to 40°C
2510-23-8
MFCD02177459
3-ethylnyl pyridine
C#CC1=CN=CC=C1
103.124
103.12

Chemical Identifiers

2510-23-8
103.124
CLRPXACRDTXENY-UHFFFAOYSA-N
186003
C#CC1=CN=CC=C1
C7H5N
MFCD02177459
3-ethylnyl pyridine
3-ethynylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

Documents