Learn More
3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5346503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Ethynylpyridine | |
39°C to 40°C | |
57°C (135°F) | |
MFCD02177459 | |
UN1325 | |
CLRPXACRDTXENY-UHFFFAOYSA-N | |
3-ethynylpyridine | |
186003 | |
97% |
2510-23-8 | |
83°C to 84°C (30 mmHg) | |
C7H5N | |
1 g | |
pyridine, 3-ethynyl, 3-pyridylacetylene, 3-ethynyl-pyridine, pyridine, 3-ethynyl-6ci, 7ci, 8ci, 9ci, 3-ethylnylpyridine, 3-ethynyl pyridine, meta-ethynylpyridine, 3-ethylnyl pyridine, 3-pyridyl acetylene | |
C#CC1=CN=CC=C1 | |
103.124 | |
103.12 |
Chemical Identifiers
2510-23-8 | |
103.124 | |
CLRPXACRDTXENY-UHFFFAOYSA-N | |
186003 | |
C#CC1=CN=CC=C1 |
C7H5N | |
MFCD02177459 | |
pyridine, 3-ethynyl, 3-pyridylacetylene, 3-ethynyl-pyridine, pyridine, 3-ethynyl-6ci, 7ci, 8ci, 9ci, 3-ethylnylpyridine, 3-ethynyl pyridine, meta-ethynylpyridine, 3-ethylnyl pyridine, 3-pyridyl acetylene | |
3-ethynylpyridine |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Hazard Class: 4.1; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only