Please login to your online account to display your discounted pricing

3-Ethynylpyridine, 96%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398070010

 View more versions of this product

Catalog No. AC398070010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Low Melting Solid
Assay ≥95%
Formula Weight 103.12
Color Colorless to brownish
Quantity 1g
CAS Min % 95.0
CAS Max % 100.0
Storage Information Product darkens on storage
Chemical Name or Material 3-Ethynylpyridine
Assay Percent Range 96%
Molecular Formula C7H5N
MDL Number MFCD02177459
CAS 2510-23-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable solid.
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Ethynylpyridine
CAS 2510-23-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H5N
Formula Weight 103.12
MDL Number MFCD02177459
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable solid.

SDS & Safety Documents