missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethyltoluene, TCI America™

Manufacturer:  TCI America E01855ML

 View more versions of this product

Catalog No. E01855ML



Specifications

Specifications

3-Ethyltoluene
C9H12
3295
ZLCSFXXPPANWQY-UHFFFAOYSA-N
1-ethyl-3-methylbenzene
12100
120.20
≥98.0% (GC)
Yellow
620-14-4
MFCD00009259
3-Methylethylbenzene
CCC1=CC=CC(=C1)C
120.195
CHEBI:77512
Clear Liquid at 20°C
160°C
5mL

Chemical Identifiers

620-14-4
120.195
ZLCSFXXPPANWQY-UHFFFAOYSA-N
12100
1-ethyl-3-methylbenzene
C9H12
MFCD00009259
3-Methylethylbenzene
CHEBI:77512
CCC1=CC=CC(=C1)C
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0015

RTECSNumber : DA0712000

TSCA : No

Documents