Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Ethylthio)propanol, 97%

$197.39 - $197.39

Chemical Identifiers

Chemical Name or Material 3-(Ethylthio)propanol
CAS 18721-61-4
Molecular Formula CH3CH2S(CH2)3OH
Formula Weight 120.21
MDL Number MFCD00236041
Beilstein 1732293
Synonym Ethyl 3-hydroxypropyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2271014

  SDS
alfa aesar™
B2271014
25g Each for $197.39
Description & Specifications

Specifications

Formula Weight 120.21
Flash Point 75°C (167°F)
Synonym Ethyl 3-hydroxypropyl sulfide
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 227° to 228°C
Molecular Formula CH3CH2S(CH2)3OH
Quantity 25g
Beilstein 1732293
MDL Number MFCD00236041
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 18721-61-4
Assay 97%
Chemical Name or Material 3-(Ethylthio)propanol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.