missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethylrhodanine, TCI America™

Manufacturer:  TCI America E017425G

 View more versions of this product

Catalog No. E017425G



Specifications

Specifications

3-Ethylrhodanine
Yellow
25g
C5H7NOS2
3-ethylrhodanine, n-ethylrhodanine, 4-thiazolidinone, 3-ethyl-2-thioxo, rhodanine, 3-ethyl, 3-ethyl-2-thioxothiazolidin-4-one, 3-ethylrodanin, 3-ethyl-2-thioxo-4-thiazolidinone, 3-ethylrhodamine, 3-ethyl-2-thioxo-1,3-thiazolidin-4-one, unii-42klq911ht
CCN1C(=O)CSC1=S
161.24
161.24
≥98.0% (GC)
128°C
37°C
7648-01-3
MFCD00051339
UPCYEFFISUGBRW-UHFFFAOYSA-N
3-ethyl-2-sulfanylidene-1,3-thiazolidin-4-one
82098
Crystal-Lump at 20°C

Chemical Identifiers

7648-01-3
161.24
UPCYEFFISUGBRW-UHFFFAOYSA-N
82098
CCN1C(=O)CSC1=S
C5H7NOS2
MFCD00051339
3-ethylrhodanine, n-ethylrhodanine, 4-thiazolidinone, 3-ethyl-2-thioxo, rhodanine, 3-ethyl, 3-ethyl-2-thioxothiazolidin-4-one, 3-ethylrodanin, 3-ethyl-2-thioxo-4-thiazolidinone, 3-ethylrhodamine, 3-ethyl-2-thioxo-1,3-thiazolidin-4-one, unii-42klq911ht
3-ethyl-2-sulfanylidene-1,3-thiazolidin-4-one
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0222

RTECSNumber : VI8092000

TSCA : Yes

Recommended Storage : Refrigerator

Documents