missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethylpyridine, TCI America™

Manufacturer:  TCI America E016925ML

 View more versions of this product

Catalog No. E016925ML



Specifications

Specifications

3-Ethylpyridine
Yellow
536-78-7
MFCD00006413
pyridine, 3-ethyl, 3-ethyl pyridine, beta-lutidine, beta-ethylpyridine, 3-ethyl-pyridine, 5-ethylpyridine, unii-a25i3ez88v, lutidine, beta, 3ethylpyridine, fema no. 3394
CCC1=CN=CC=C1
107.156
107.16
≥98.0% (GC)
165°C
25mL
C7H9N
1993
MFEIKQPHQINPRI-UHFFFAOYSA-N
3-ethylpyridine
10823
Clear Liquid at 20°C

Chemical Identifiers

536-78-7
107.156
MFEIKQPHQINPRI-UHFFFAOYSA-N
10823
CCC1=CN=CC=C1
C7H9N
MFCD00006413
pyridine, 3-ethyl, 3-ethyl pyridine, beta-lutidine, beta-ethylpyridine, 3-ethyl-pyridine, 5-ethylpyridine, unii-a25i3ez88v, lutidine, beta, 3ethylpyridine, fema no. 3394
3-ethylpyridine
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0222

TSCA : Yes

Documents