Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethylphenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 3-Ethylphenyl isocyanate
CAS 23138-58-1
Molecular Formula C9H9NO
Formula Weight 147.18g/mol
MDL Number MFCD00013870
UN Number UN2206
Beilstein 386718
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1269903

 
alfa aesar™
L12699-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1269906

 
alfa aesar™
L12699-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 106° to 107°C (40mmHg)
Quantity 1g
Flash Point 85°C (185°F)
Odor Pungent
Chemical Name or Material 3-Ethylphenyl isocyanate
CAS 23138-58-1
Assay 97%
Molecular Formula C9H9NO
Formula Weight 147.18g/mol
MDL Number MFCD00013870
UN Number UN2206
Beilstein 386718
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C