Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethylphenol, 95%

Chemical Identifiers

Chemical Name or Material 3-Ethylphenol
CAS 620-17-7
Molecular Formula C8H10O
Formula Weight 122.17
MDL Number MFCD00002311
UN Number UN3145
Beilstein 1857002
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2347009

 
alfa aesar™
B2347009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2347018

 
alfa aesar™
B2347018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2347030

 
alfa aesar™
B2347030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Chemical Name or Material 3-Ethylphenol
TSCA TSCA
Assay 95%
Molecular Formula C8H10O
MDL Number MFCD00002311
CAS 620-17-7
UN Number UN3145
Beilstein 1857002
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.
Flash Point 94°C (202°F)
Boiling Point 218°C
Melting Point −4°C
Quantity 10g

Applications
It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.