Learn More
3-Ethylamino-4-methylphenol, tech. 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1276806
Description
3-Ethylamino-4-methylphenol was used in the synthesis of rhodol by undergoing condensation reaction with 2-(2,4-dihydroxybenzoyl)benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Ethylamino-4-methylphenol was used in the synthesis of rhodol by undergoing condensation reaction with 2-(2,4-dihydroxybenzoyl)benzoic acid.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
3-Ethylamino-4-methylphenol | |
84°C to 92°C | |
C9H13NO | |
5 g | |
3-ethylamino-4-methylphenol, 3-ethylamino-p-cresol, phenol, 3-ethylamino-4-methyl, ccris 4639, 3-ethylamino-4-methyl-phenol, unii-8002qme89h, 3-ethylamino-4-cresol, 3-ethylamino p-cresol, dsstox_cid_5269 | |
CTGSQPRDMHCIMM-UHFFFAOYSA-N | |
3-(ethylamino)-4-methylphenol | |
8428 | |
90% |
120-37-6 | |
135°C to 140°C (3 mmHg) | |
MFCD00002382 | |
3241548 | |
Insoluble in water. | |
CCNC1=CC(O)=CC=C1C | |
151.21 | |
151.21 | |
Technical |
Chemical Identifiers
120-37-6 | |
151.21 | |
CTGSQPRDMHCIMM-UHFFFAOYSA-N | |
8428 | |
CCNC1=CC(O)=CC=C1C |
C9H13NO | |
MFCD00002382 | |
3-ethylamino-4-methylphenol, 3-ethylamino-p-cresol, phenol, 3-ethylamino-4-methyl, ccris 4639, 3-ethylamino-4-methyl-phenol, unii-8002qme89h, 3-ethylamino-4-cresol, 3-ethylamino p-cresol, dsstox_cid_5269 | |
3-(ethylamino)-4-methylphenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-391-0
RTECSNumber : SL4105000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only