Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-5-hexen-3-ol, 97%

$267.84 - $940.50

Chemical Identifiers

Chemical Name or Material 3-Ethyl-5-hexen-3-ol
CAS 1907-46-6
Molecular Formula C8H16O
Formula Weight 128.21
MDL Number MFCD00021834
Synonym Allyl diethyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2042214

  SDS
alfa aesar™
B2042214
25g Each for $267.84

AAB2042222

  SDS
alfa aesar™
B2042222
100g Each for $940.50
Description & Specifications

Specifications

Formula Weight 128.21
Molecular Formula C8H16O
MDL Number MFCD00021834
CAS 1907-46-6
Synonym Allyl diethyl carbinol
Quantity 25g
Chemical Name or Material 3-Ethyl-5-hexen-3-ol
Assay 97%