Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-4-octanone, 98%

$114.36 - $460.24

Chemical Identifiers

Chemical Name or Material 3-Ethyl-4-octanone
CAS 19781-29-4
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00048806
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2017506

 
alfa aesar™
B2017506
5g Each for $114.36

AAB2017514

 
alfa aesar™
B2017514
25g Each for $460.24
Description & Specifications

Specifications

Formula Weight 156.27
Chemical Name or Material 3-Ethyl-4-octanone
Assay 98%
Molecular Formula C10H20O
MDL Number MFCD00048806
CAS 19781-29-4
Quantity 5g