Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-4-methylpyridine, 95%

$175.47 - $714.23

Chemical Identifiers

Chemical Name or Material 3-Ethyl-4-methylpyridine
CAS 529-21-5
Molecular Formula C8H11N
Formula Weight 121.18
MDL Number MFCD00014638
Synonym 3-Ethyl-4-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2140606

  SDS
alfa aesar™
B2140606
5g Each for $714.23

AAB2140603

  SDS
alfa aesar™
B2140603
1g Each for $175.47
Description & Specifications

Specifications

Formula Weight 121.18
Chemical Name or Material 3-Ethyl-4-methylpyridine
TSCA TSCA
Assay 95%
Molecular Formula C8H11N
MDL Number MFCD00014638
CAS 529-21-5
Synonym 3-Ethyl-4-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 195° to 196°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.