missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-4-methyl-3-pyrrolin-2-one, 98%

Manufacturer:  Alfa Aesar™ H5506503

 View more versions of this product

Catalog No. AAH5506503



Description

Description

Applications
3-Ethyl-4-methyl-3-pyrrolin-2-one is used as an intermediate for the manufacture of medicines. It is used for the preparation of Glimepiride (G410150), and its intermediates and as antidiabetic agents.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

3-Ethyl-4-methyl-3-pyrrolin-2-one
1g
C7H11NO
1,5-dihydro-3-ethyl-4-methyl-2h-pyrrol-2-one
YCTNTSVMJWIYTQ-UHFFFAOYSA-N
4-ethyl-3-methyl-1,2-dihydropyrrol-5-one
854146
98%
100°C to 104°C
766-36-9
MFCD00173861
Slightly soluble in water.
CCC1=C(CNC1=O)C
125.171
125.17

Chemical Identifiers

766-36-9
125.171
YCTNTSVMJWIYTQ-UHFFFAOYSA-N
854146
CCC1=C(CNC1=O)C
C7H11NO
MFCD00173861
1,5-dihydro-3-ethyl-4-methyl-2h-pyrrol-2-one
4-ethyl-3-methyl-1,2-dihydropyrrol-5-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents