missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethyl-3-pentanol, TCI America™

Manufacturer:  TCI America E015710ML

 View more versions of this product

Catalog No. E015710ML



Specifications

Specifications

3-Ethyl-3-pentanol
Colorless
597-49-9
MFCD00004484
3-ethyl-3-pentanol, triethylcarbinol, 3-pentanol, 3-ethyl, triethylmethanol, triethylmethyl alcohol, tert-heptanol, 3-aethyl-pentanol-3, 3-aethyl-pentanol-3 german, 3-ethyl-3-hydroxypentane, ethyl-3 pentanol-3
CCC(CC)(CC)O
116.204
116.20
≥99.0% (GC)
143°C
10mL
C7H16O
1987
XKIRHOWVQWCYBT-UHFFFAOYSA-N
3-ethylpentan-3-ol
11702
Clear Liquid at 20°C

Chemical Identifiers

597-49-9
116.204
XKIRHOWVQWCYBT-UHFFFAOYSA-N
11702
CCC(CC)(CC)O
C7H16O
MFCD00004484
3-ethyl-3-pentanol, triethylcarbinol, 3-pentanol, 3-ethyl, triethylmethanol, triethylmethyl alcohol, tert-heptanol, 3-aethyl-pentanol-3, 3-aethyl-pentanol-3 german, 3-ethyl-3-hydroxypentane, ethyl-3 pentanol-3
3-ethylpentan-3-ol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : SA6780000

TSCA : Yes

Documents