missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethyl-3-oxetanemethanol, TCI America™

Manufacturer:  TCI America E0546100G

 View more versions of this product

Catalog No. E0546100G



Specifications

Specifications

3-Ethyl-3-oxetanemethanol
Colorless
3047-32-3
MFCD00190143
UNMJLQGKEDTEKJ-UHFFFAOYSA-N
(3-ethyloxetan-3-yl)methanol
76444
Clear Liquid at 20°C
110°C
100g
C6H12O2
3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane
CCC1(COC1)CO
116.16
116.16
≥96.0% (GC)

Chemical Identifiers

3047-32-3
116.16
UNMJLQGKEDTEKJ-UHFFFAOYSA-N
76444
CCC1(COC1)CO
C6H12O2
MFCD00190143
3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane
(3-ethyloxetan-3-yl)methanol
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6621

TSCA : Yes

Documents