missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-3-oxetanemethanol, 97%

Manufacturer:  Alfa Aesar™ H5349714

 View more versions of this product

Catalog No. AAH5349714



Description

Description

Applications
3-Ethyl-3-oxetanemethanol is used as pharmaceutical intermediate.

Solubility
Miscible in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

3-Ethyl-3-oxetanemethanol
108°C (226°F)
96°C (4mmHg)
3047-32-3
MFCD00190143
Miscible in water.
CCC1(COC1)CO
116.16
116.16
1.019
1.453
25g
C6H12O2
2-hydroxymethyl-2-ethyl-1,3-epoxypropane
UNMJLQGKEDTEKJ-UHFFFAOYSA-N
(3-ethyloxetan-3-yl)methanol
76444
97%

Chemical Identifiers

3047-32-3
116.16
UNMJLQGKEDTEKJ-UHFFFAOYSA-N
76444
CCC1(COC1)CO
C6H12O2
MFCD00190143
2-hydroxymethyl-2-ethyl-1,3-epoxypropane
(3-ethyloxetan-3-yl)methanol
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 221-254-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents