missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethyl-3-oxetanemethanol, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC377700250

 View more versions of this product

Catalog No. AC377700250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Ethyl-3-oxetanemethanol
25g
95.0
100.0
Glass bottle
Liquid
96%
116.16
Clear colorless
3047-32-3
Authentic
1.019g/mL
1.4520 to 1.4540 (20°C, 589nm)
≥95%
C6H12O2
MFCD00190143

Chemical Identifiers

3047-32-3
100.0
C6H12O2
MFCD00190143
≥95%
95.0
96%
116.16
3-Ethyl-3-oxetanemethanol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.