Please login to your online account to display your discounted pricing

3-Ethyl-3-oxetanemethanol, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC377700050

 View more versions of this product

Catalog No. AC377700050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 116.16
Quantity 5g
Color Clear colorless
Density 1.019g/mL
CAS Min % 95.0
CAS Max % 100.0
Refractive Index 1.4520 to 1.4540 (20°C, 589nm)
Molecular Formula C6H12O2
MDL Number MFCD00190143
Assay Percent Range 96%
CAS 3047-32-3
Chemical Name or Material 3-Ethyl-3-oxetanemethanol
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 3-Ethyl-3-oxetanemethanol
CAS 3047-32-3
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C6H12O2
Formula Weight 116.16
MDL Number MFCD00190143
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents