Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-2-methylbenzoxazolium iodide, 95%

$216.97 - $741.32

Chemical Identifiers

Chemical Name or Material 3-Ethyl-2-methylbenzoxazolium iodide
CAS 5260-37-7
Molecular Formula C10H12INO
Formula Weight 289.12
MDL Number MFCD00044141
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1877822

  SDS
alfa aesar™
A1877822
100g Each for $741.32

AAA1877814

  SDS
alfa aesar™
A1877814
25g Each for $216.97

AAA1877806

  SDS
alfa aesar™
A1877806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-Ethyl-2-methylbenzoxazolium iodide
CAS 5260-37-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DM5285000
TSCA TSCA
Assay 95%
Molecular Formula C10H12INO
Formula Weight 289.12
MDL Number MFCD00044141
Melting Point 196° to 198°C (dec.)
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.