Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-2-methyl-2-heptanol, 97%

$134.24 - $518.32

Chemical Identifiers

Chemical Name or Material 3-Ethyl-2-methyl-2-heptanol
CAS 19780-59-7
Molecular Formula C10H22O
Formula Weight 158.28
MDL Number MFCD00046575
Synonym Dimethyl 3-heptyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2097906

  SDS
alfa aesar™
B2097906
5g Each for $134.24

AAB2097914

  SDS
alfa aesar™
B2097914
25g Each for $518.32
Description & Specifications

Specifications

Formula Weight 158.28
CAS 19780-59-7
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Dimethyl 3-heptyl carbinol
MDL Number MFCD00046575
Chemical Name or Material 3-Ethyl-2-methyl-2-heptanol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Molecular Formula C10H22O
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.