missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethyl-2-hydroxy-2-cyclopenten-1-one, TCI America™

Manufacturer:  TCI America E079225G

 View more versions of this product

Catalog No. E079225G



Specifications

Specifications

3-Ethyl-2-hydroxy-2-cyclopenten-1-one
White-Yellow
25g
C7H10O2
JHWFWLUAUPZUCP-UHFFFAOYSA-N
3-ethyl-2-hydroxycyclopent-2-en-1-one
62752
Crystal-Powder at 20°C
75°C
42°C
21835-01-8
MFCD00012391
CCC1=C(C(=O)CC1)O
126.155
126.16
≥97.0% (GC)

Chemical Identifiers

21835-01-8
126.155
JHWFWLUAUPZUCP-UHFFFAOYSA-N
3-ethyl-2-hydroxycyclopent-2-en-1-one
C7H10O2
MFCD00012391
62752
CCC1=C(C(=O)CC1)O
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Freezer

Documents