Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-2-heptanol, erythro + threo, 98%

$80.04 - $318.64

Chemical Identifiers

Chemical Name or Material 3-Ethyl-2-heptanol
CAS 19780-39-3
Name Note erythro + threo
Molecular Formula C9H20O
Formula Weight 144.26
MDL Number MFCD00046644
Synonym 3-Heptyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2065206

 
alfa aesar™
B2065206
5g Each for $80.04

AAB2065214

 
alfa aesar™
B2065214
25g Each for $318.64
Description & Specifications

Specifications

Formula Weight 144.26
Name Note erythro + threo
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Chemical Name or Material 3-Ethyl-2-heptanol
Assay 98%
Molecular Formula C9H20O
MDL Number MFCD00046644
CAS 19780-39-3
Synonym 3-Heptyl methyl carbinol
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.